Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mesdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030705.4 | NP_109630.1 | 1 | 1605 | ||
GenBank mRNA | AK084510.1 | - | 1 | 1551 | ||
AK089082.1 | - | 1 | 1560 | |||
AK162446.1 | - | 1 | 1492 | |||
BC018326.1 | - | 1 | 614 | 4018100 |