Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030709.2 | NP_109634.2 | 3 | 277 | ||
GenBank mRNA | AB016229.1 | - | 3 | 285 | ||
AB016230.1 | - | 3 | 277 | |||
AB041037.1 | - | 3 | 234 | |||
AK029714.1 | - | 4 | 1124 | |||
BC132151.1 | - | 2 | 223 | 40130428 | ||
BC145027.1 | - | 2 | 223 | 9053575 |