Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl1xr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030732.3 | NP_109657.2 | 4, 5 | 627 | ||
GenBank mRNA | AF268195.1 | - | 4, 5 | 408 | ||
AK029595.1 | - | 4, 5 | 426 | |||
AK033347.1 | - | 4, 5 | 497 | |||
AK036064.1 | - | 5, 6 | 738 | |||
AK136255.1 | - | 4, 5 | 449 |