Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030750.3 | NP_109675.1 | 3 | 1280 | ||
GenBank mRNA | AF247177.1 | - | 3 | 1230 | ||
AF415215.1 | - | 3 | 1114 | |||
AK149860.1 | - | 3 | 1281 | |||
AK154116.1 | - | 4 | 922 | |||
BC026814.1 | - | 3 | 1216 | |||
BC037592.1 | - | 3 | 1233 | 5004335 |