Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorcs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030889.2 | NP_112151.2 | 2, 3 | 650 | ||
GenBank mRNA | AB093294.1 | - | 2, 3 | 635 | ||
AK047881.1 | - | 2, 3 | 651 | |||
AK053853.1 | - | 2, 3 | 620 | |||
AY004316.1 | - | 2, 3 | 619 | |||
BC138873.1 | - | 2, 3 | 576 | 8861895 | ||
BC138875.1 | - | 2, 3 | 576 | 8861897 |