Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ide | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031156.3 | NP_112419.3 | 3, 4 | 695 | ||
GenBank mRNA | AJ278422.1 | - | 3, 4 | 715 | ||
AK004972.2 | - | 3, 4 | 695 | |||
AK014703.1 | - | 615 | ||||
AK128986.1 | - | 3, 4 | 712 | |||
BC041675.1 | - | 3, 4 | 727 | 4951971 |