Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030677.2 | NP_109602.2 | 2 | 854 | ||
GenBank mRNA | AK131940.1 | - | 2 | 731 | ||
BC010823.1 | - | 2 | 724 | 4218623 | ||
BC034335.1 | - | 2 | 701 | 5038341 | ||
BC039658.1 | - | 2 | 799 | 4987478 | ||
BC054848.1 | - | 2 | 677 | 6529267 | ||
BC066825.1 | - | 1 | 2951 |