Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rai14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166408.1 | NP_001159880.1 | 9 | 857 | ||
NM_030690.3 | NP_109615.1 | 9 | 943 | |||
GenBank mRNA | AF202315.1 | - | 8 | 730 | ||
AF274866.1 | - | 9 | 922 | |||
AK129333.1 | - | 9 | 793 | |||
AK141233.1 | - | 5 | 443 | |||
AK160119.1 | - | 9 | 853 | |||
BC052458.1 | - | 9 | 943 | 6830677 |