Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030691.1 | NP_109616.1 | 6 | 2009 | ||
GenBank mRNA | AJ278575.1 | - | 6 | 2009 | ||
AK040569.1 | - | 6 | 1990 | |||
AK152186.1 | - | 6 | 1998 | |||
AK152580.1 | - | 1 | 1005 | |||
AK156382.1 | - | 6 | 1947 | |||
AK163058.1 | - | 6 | 1990 | |||
BC057951.1 | - | 6 | 2006 | 5341788 |