Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007118.3 | NP_009049.2 | 3 | 346 | ||
NR_134469.1 | 3 | 346 | ||||
XM_011514107.2 | XP_011512409.1 | 3 | 921 | |||
XM_011514108.1 | XP_011512410.1 | 3 | 220 | |||
XM_011514109.2 | XP_011512411.1 | 3 | 3998 | |||
XM_011514110.2 | XP_011512412.1 | 3 | 456 | |||
XM_017009801.1 | XP_016865290.1 | 3 | 698 | |||
XM_017009802.1 | XP_016865291.1 | 3 | 698 | |||
GenBank mRNA | AF091395.1 | - | 3 | 211 | ||
AK304786.1 | - | 3 | 325 | |||
BC035585.1 | - | 3 | 271 | |||
BF058117.1 | - | 3 | 346 | |||
U42390.1 | - | 3 | 211 |