Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038653.1 | NP_001033742.1 | 6 | 2107 | ||
NM_001038654.1 | NP_001033743.1 | 5 | 2012 | |||
NM_030696.3 | NP_109621.1 | 6 | 2119 | |||
GenBank mRNA | AF178954.1 | - | 4 | 1907 | ||
AK141381.1 | - | 5 | 2013 | |||
AK149707.1 | - | 6 | 2110 | |||
AK149983.1 | - | 6 | 2110 | |||
AK165596.1 | - | 5 | 1956 | |||
AK166048.1 | - | 2095 | ||||
AK169917.1 | - | 6 | 2069 | |||
BC046525.1 | - | 6 | 2120 | 5319754 |