Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcrls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030707.3 | NP_109632.2 | 3 | 216 | ||
GenBank mRNA | AB028174.1 | - | 3 | 247 | ||
AF302046.1 | - | 3 | 285 | |||
AF329486.1 | - | 3 | 217 | |||
AK034125.1 | - | 3 | 217 | |||
AK076123.1 | - | 3 | 250 | |||
AY506556.1 | - | 3 | 256 | |||
AY506557.1 | - | 3 | 256 | |||
BC016551.1 | - | 3 | 259 | 3602803 |