Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030715.3 | NP_109640.1 | 11 | 2349 | ||
GenBank mRNA | AB027128.1 | - | 11 | 2375 | ||
AB037184.1 | - | 4 | 1127 | |||
AK036296.1 | - | 11 | 2350 | |||
BC049159.1 | - | 11 | 2319 |