Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030724.3 | NP_109649.1 | 2 | 495 | ||
GenBank mRNA | AF236636.1 | - | 2 | 500 | ||
AK051894.1 | - | 2 | 265 | |||
AK053745.1 | - | 2 | 455 | |||
AK077517.1 | - | 2 | 432 | |||
AK082837.1 | - | 2 | 458 | |||
AK155939.1 | - | 2 | 459 | |||
AK168318.1 | - | 2 | 473 | |||
BC023789.1 | - | 2 | 459 | 5352192 |