Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166844.1 | NP_001160316.1 | 1 | 378 |
Gene Symbol: Gm16456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC107032.2 | - | 1 | 377 | ||
BC134368.1 | - | 1 | 377 |
Gene Symbol: Gm4141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167163.1 | NP_001160635.1 | 1 | 378 |
Gene Symbol: Gm4201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167162.1 | NP_001160634.1 | 1 | 378 |
Gene Symbol: Gm4498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167151.1 | NP_001160623.1 | 1 | 378 |
Gene Symbol: Gm8453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167150.1 | NP_001160622.1 | 1 | 378 |
Gene Symbol: Gm8693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167154.1 | NP_001160626.1 | 1 | 378 |
Gene Symbol: Vmn1r104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166738.1 | NP_001160210.1 | 1 | 378 |
Gene Symbol: Vmn1r107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166759.1 | NP_001160231.1 | 1 | 378 |
Gene Symbol: Vmn1r111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166746.1 | NP_001160218.1 | 1 | 378 |
Gene Symbol: Vmn1r112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166847.1 | NP_001160319.1 | 1 | 378 |
Gene Symbol: Vmn1r116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166744.1 | NP_001160216.1 | 1 | 378 |
Gene Symbol: Vmn1r118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166742.1 | NP_001160214.1 | 1 | 378 |
Gene Symbol: Vmn1r123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166707.1 | NP_001160179.1 | 1 | 378 |
Gene Symbol: Vmn1r125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166740.1 | NP_001160212.1 | 1 | 378 |
Gene Symbol: Vmn1r128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166739.1 | NP_001160211.1 | 1 | 378 |
Gene Symbol: Vmn1r130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166848.1 | NP_001160320.1 | 1 | 378 |
Gene Symbol: Vmn1r131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166839.1 | NP_001160311.1 | 1 | 378 |
Gene Symbol: Vmn1r132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122682.1 | NP_001116154.1 | 1 | 1725 |
Gene Symbol: Vmn1r135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166747.1 | NP_001160219.1 | 1 | 378 |
Gene Symbol: Vmn1r137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166849.1 | NP_001160321.1 | 1 | 378 |
Gene Symbol: Vmn1r138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167169.1 | NP_001160641.1 | 1 | 378 |
Gene Symbol: Vmn1r139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166748.1 | NP_001160220.1 | 1 | 378 |
Gene Symbol: Vmn1r143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166750.1 | NP_001160222.1 | 1 | 378 |
Gene Symbol: Vmn1r148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030736.2 | NP_109661.1 | 1 | 1730 | ||
GenBank mRNA | AF324872.1 | - | 1 | 1732 |
Gene Symbol: Vmn1r152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166752.1 | NP_001160224.1 | 1 | 378 |
Gene Symbol: Vmn1r155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166753.1 | NP_001160225.1 | 1 | 378 |
Gene Symbol: Vmn1r157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166754.1 | NP_001160226.1 | 1 | 378 |
Gene Symbol: Vmn1r160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166724.1 | NP_001160196.1 | 1 | 378 |
Gene Symbol: Vmn1r163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166755.1 | NP_001160227.1 | 1 | 378 |
Gene Symbol: Vmn1r165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166850.1 | NP_001160322.1 | 1 | 378 |
Gene Symbol: Vmn1r166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167168.1 | NP_001160640.1 | 1 | 378 |
Gene Symbol: Vmn1r95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167538.1 | NP_001161010.1 | 1 | 378 |