Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030749.2 | NP_109674.2 | 4 | 498 | ||
GenBank mRNA | AJ297884.1 | - | 4 | 439 | ||
AK170415.1 | - | 4 | 451 | |||
BC016119.1 | - | 4 | 475 | 4489217 | ||
BC016466.1 | - | 4 | 461 | 3582798 | ||
CO428760.1 | - | 1 | 41 | |||
CX219879.1 | - | 4 | 498 |