Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorcs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030889.2 | NP_112151.2 | 27 | 4420 | ||
GenBank mRNA | AB093294.1 | - | 27 | 4405 | ||
AK160976.1 | - | 8 | 1690 | |||
AY004316.1 | - | 27 | 4389 | |||
BC022668.1 | - | 1 | 41 | |||
BC138873.1 | - | 27 | 4346 | 8861895 | ||
BC138875.1 | - | 27 | 4346 | 8861897 |