Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000440.2 | NP_000431.2 | 1 | 335 | ||
XM_017009572.1 | XP_016865061.1 | 1 | 335 | |||
GenBank mRNA | AB593141.1 | - | 1 | 335 | ||
BC035909.1 | - | 1 | 322 | 5492027 | ||
BC144044.1 | - | 1 | 262 | |||
M26061.1 | - | 1 | 335 |