Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HSPA8 SIRNA Assays ›
Gene Symbol:
Hspa8
Gene Name:
heat shock protein 8
Gene Aliases:
2410008N15Rik, Hsc70, Hsc71, Hsc73, Hsp73, Hspa10
Chromosome Location:
Chr.9: 40801273 - 40805199 on Build GRCm38
UniGene ID:
Mm.290774
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
84860
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hspa8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031165.4
NP_112442.2
2
164
GenBank mRNA
AK004608.1
-
2
162
AK035286.1
-
2
162
AK075935.1
-
2
161
AK081701.1
-
2
162
AK098074.1
-
2
161
AK145579.1
-
2
158
AK146547.1
-
2
165
AK146708.1
-
2
159
AK146985.1
-
2
162
AK147864.1
-
2
162
AK149954.1
-
2
159
AK150498.1
-
2
156
AK150701.1
-
2
166
AK150958.1
-
2
159
AK151041.1
-
2
162
AK151065.1
-
2
162
AK151127.1
-
2
165
AK151287.1
-
2
162
AK151435.1
-
2
162
AK151516.1
-
2
168
AK151537.1
-
2
159
AK151741.1
-
2
162
AK151775.1
-
2
164
AK151808.1
-
2
165
AK151865.1
-
2
162
AK151892.1
-
2
164
AK151948.1
-
2
165
AK151997.1
-
2
162
AK152315.1
-
2
165
AK152598.1
-
2
162
AK152697.1
-
2
162
AK152703.1
-
2
162
AK152768.1
-
2
165
AK152803.1
-
2
165
AK153032.1
-
2
164
AK153834.1
-
2
162
AK157831.1
-
2
162
AK159479.1
-
2
162
AK162775.1
-
2
162
AK163987.1
-
2
162
AK164000.1
-
2
162
AK166545.1
-
2
162
AK166643.1
-
2
176
AK166721.1
-
2
159
AK166767.1
-
2
162
AK166776.1
-
2
165
AK166808.1
-
2
162
AK166830.1
-
2
160
AK166846.1
-
2
162
AK166861.1
-
2
165
AK166873.1
-
2
162
AK166908.1
-
2
165
AK166910.1
-
2
162
AK166913.1
-
2
162
AK166933.1
-
2
162
AK167043.1
-
2
164
AK167121.1
-
2
165
AK167122.1
-
2
162
AK167134.1
-
2
160
AK167163.1
-
2
164
AK167218.1
-
2
162
AK167229.1
-
2
163
AK167254.1
-
2
162
AK167414.1
-
2
162
AK167845.1
-
2
165
AK167910.1
-
2
165
AK168435.1
-
2
165
AK168492.1
-
2
164
AK168519.1
-
2
162
AK168542.1
-
2
164
AK168577.1
-
2
162
AK168625.1
-
2
162
AK168654.1
-
2
162
AK168711.1
-
2
162
AK168750.1
-
2
165
AK168776.1
-
2
165
AK168887.1
-
2
164
AK168934.1
-
2
165
AK169093.1
-
2
162
AK169179.1
-
2
162
AK169236.1
-
2
162
AK169293.1
-
2
176
AK169329.1
-
2
165
AK169594.1
-
2
161
AK171434.1
-
2
163
AK190650.1
-
2
150
AK196709.1
-
2
150
AK196874.1
-
2
150
AK197713.1
-
150
AK197877.1
-
2
150
AK197978.1
-
2
150
AK201032.1
-
2
150
AK201241.1
-
2
133
AK202184.1
-
2
152
AK202337.1
-
2
150
AK205346.1
-
2
150
AK206185.1
-
2
150
AK207066.1
-
1
35
AK207762.1
-
2
150
AK210894.1
-
2
150
AK210977.1
-
2
150
AK214010.1
-
2
150
BC006722.1
-
2
186
3711399
BC066191.1
-
2
118
30467052
BC085486.1
-
2
150
30540914
BC089322.1
-
2
143
30538640
BC089457.1
-
2
121
30628505
BC094900.1
-
2
135
30063483
BC106193.1
-
2
146
6329283
M19141.1
-
2
148
U27129.1
-
2
113
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
protein folding
RNA splicing
protein refolding
negative regulation of transcription, DNA-templated
ATP metabolic process
positive regulation of mRNA splicing, via spliceosome
chaperone mediated protein folding requiring cofactor
regulation of cell cycle
chaperone-mediated protein folding
clathrin coat disassembly
negative regulation of fibril organization
nucleotide binding
G-protein coupled receptor binding
receptor binding
protein binding
ATP binding
ATPase activity
enzyme binding
MHC class II protein complex binding
heat shock protein binding
ubiquitin protein ligase binding
ATPase activity, coupled
ADP binding
poly(A) RNA binding
unfolded protein binding
C3HC4-type RING finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline