Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnmb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031169.4 | NP_112446.2 | 3 | 493 | ||
GenBank mRNA | AF020711.1 | - | 3 | 493 | ||
AJ001291.1 | - | 3 | 235 | |||
AK038987.1 | - | 3 | 221 | |||
BC013338.1 | - | 3 | 284 | 3988005 |