Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031182.2 | NP_112459.1 | 2 | 388 | ||
GenBank mRNA | AF161262.1 | - | 2 | 388 | ||
AK053091.1 | - | 2 | 180 | |||
AK166424.1 | - | 2 | 527 | |||
BC047270.1 | - | 2 | 295 | 6438955 | ||
BC054777.1 | - | 2 | 279 | 6808430 |