Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PECAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000442.4 | NP_000433.4 | 3 | 452 | ||
XM_005276880.1 | XP_005276937.1 | 3 | 738 | |||
XM_005276881.1 | XP_005276938.1 | 3 | 738 | |||
XM_005276882.1 | XP_005276939.1 | 3 | 738 | |||
XM_005276883.2 | XP_005276940.1 | 3 | 738 | |||
XM_011524889.2 | XP_011523191.1 | 4 | 468 | |||
XM_011524890.1 | XP_011523192.1 | 4 | 417 | |||
XM_017024738.1 | XP_016880227.1 | 4 | 361 | |||
XM_017024739.1 | XP_016880228.1 | 3 | 738 | |||
XM_017024740.1 | XP_016880229.1 | 4 | 431 | |||
XM_017024741.1 | XP_016880230.1 | 3 | 738 | |||
GenBank mRNA | AF281287.1 | - | 3 | 322 | ||
AF281288.1 | - | 3 | 322 | |||
AF281289.1 | - | 3 | 322 | |||
AF281290.1 | - | 3 | 322 | |||
AF281291.1 | - | 3 | 322 | |||
AF281292.1 | - | 3 | 322 | |||
AF281293.1 | - | 3 | 322 | |||
AF281294.1 | - | 3 | 322 | |||
AF281295.1 | - | 3 | 322 | |||
AF281296.1 | - | 3 | 322 | |||
AF281297.1 | - | 3 | 322 | |||
AF281298.1 | - | 3 | 322 | |||
AF281299.1 | - | 3 | 322 | |||
AF281300.1 | - | 3 | 322 | |||
AF281301.1 | - | 3 | 322 | |||
AF393677.1 | - | 3 | 322 | |||
AF393678.1 | - | 3 | 322 | |||
AK290692.1 | - | 3 | 255 | |||
AK304166.1 | - | 2 | 252 | |||
BC022512.1 | - | 3 | 432 | 4816878 | ||
BC051822.1 | - | 3 | 452 | 5768579 | ||
JQ287500.1 | - | 3 | 239 | |||
M28526.1 | - | 3 | 380 | |||
M37780.1 | - | 3 | 364 |