Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031198.3 | NP_112475.1 | 2 | 382 | ||
GenBank mRNA | AF077742.1 | - | 2 | 382 | ||
AK137141.1 | - | 2 | 347 | |||
AK156327.1 | - | 2 | 295 | |||
AK157426.1 | - | 2 | 382 | |||
AK159579.1 | - | 2 | 315 | |||
BC052832.1 | - | 2 | 345 | |||
BC064770.1 | - | 2 | 333 | |||
BC098494.1 | - | 2 | 267 | 6431607 |