Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282014.1 | NP_001268943.1 | 2 | 560 | ||
NM_001282015.1 | NP_001268944.1 | 2 | 554 | |||
NM_031202.3 | NP_112479.1 | 2 | 575 | |||
GenBank mRNA | AK148332.1 | - | 2 | 560 | ||
AK148370.1 | - | 2 | 466 | |||
AK148441.1 | - | 2 | 466 | |||
AK185619.1 | - | 1 | 272 | |||
AK190422.1 | - | 1 | 58 | |||
AK191462.1 | - | 1 | 58 | |||
AK191697.1 | - | 1 | 57 | |||
AK192907.1 | - | 1 | 56 | |||
AK192940.1 | - | 1 | 58 | |||
AK195260.1 | - | 1 | 23 | |||
AK199454.1 | - | 1 | 15 | |||
AK200238.1 | - | 1 | 58 | |||
AK201835.1 | - | 1 | 58 | |||
AK202735.1 | - | 1 | 58 | |||
AK207360.1 | - | 1 | 25 | |||
AK219128.1 | - | 1 | 272 | |||
AK219226.1 | - | 1 | 272 | |||
BC076598.1 | - | 2 | 558 | 30613975 | ||
X03687.1 | - | 2 | 460 |