Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007118.3 | NP_009049.2 | 8 | 1496 | ||
NR_134469.1 | 8 | 1496 | ||||
XM_011514107.2 | XP_011512409.1 | 8 | 2071 | |||
XM_011514108.1 | XP_011512410.1 | 8 | 1370 | |||
XM_011514109.2 | XP_011512411.1 | 8 | 5148 | |||
XM_011514110.2 | XP_011512412.1 | 8 | 1606 | |||
XM_017009801.1 | XP_016865290.1 | 8 | 1848 | |||
XM_017009802.1 | XP_016865291.1 | 8 | 1848 | |||
GenBank mRNA | AB209754.1 | - | 3 | 390 | ||
AF091395.1 | - | 8 | 1361 | |||
AK304786.1 | - | 8 | 1475 | |||
BC035585.1 | - | 8 | 1421 | |||
U42390.1 | - | 8 | 1361 |