Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031374.2 | NP_113551.2 | 1 | 90 | ||
GenBank mRNA | AF285589.1 | - | 1 | 90 | ||
AK143304.1 | - | 6 | 862 |