Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031375.4 | NP_113552.3 | 3 | 370 | ||
GenBank mRNA | AB047544.1 | - | 3 | 396 | ||
AK083330.1 | - | 2 | 763 | |||
AK144756.1 | - | 3 | 366 | |||
AK146059.1 | - | 2 | 745 | |||
AK181653.1 | - | 1 | 52 | |||
BC004034.1 | - | 357 | 3495639 |