Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031393.2 | NP_113570.2 | 3 | 470 | ||
GenBank mRNA | AB050741.1 | - | 3 | 449 | ||
AK078868.1 | - | 3 | 458 | |||
AK131162.1 | - | 3 | 427 | |||
AK154176.1 | - | 3 | 465 | |||
BC005623.1 | - | 3 | 470 | 3155535 |