Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031406.3 | NP_113583.2 | 5 | 671 | ||
GenBank mRNA | AF314957.1 | - | 4 | 537 | ||
AK154118.1 | - | 5 | 629 | |||
BC046982.1 | - | 5 | 591 | 6478046 |