Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000452.2 | NP_000443.1 | 4 | 1242 | ||
GenBank mRNA | BC130521.1 | - | 4 | 678 | 40146552 | |
BC130523.1 | - | 4 | 678 | 40146554 | ||
U10417.1 | - | 4 | 1243 |