Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QDPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000320.2 | NP_000311.2 | 7 | 1591 | ||
NM_001306140.1 | NP_001293069.1 | 6 | 1498 | |||
GenBank mRNA | M16447.1 | - | 7 | 1495 |