Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otof | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100395.1 | NP_001093865.1 | 2 | 259 | ||
NM_001286421.1 | NP_001273350.1 | 2 | 259 | |||
NM_031875.2 | NP_114081.2 | 2 | 259 | |||
GenBank mRNA | AF183183.1 | - | 2 | 229 | ||
AF183184.1 | - | 2 | 229 | |||
AK046460.1 | - | 2 | 261 | |||
AK147256.1 | - | 2 | 242 | |||
AK147434.1 | - | 2 | 242 | |||
AY586513.1 | - | 2 | 125 | |||
BB645102.1 | - | 2 | 261 | |||
BC145107.1 | - | 2 | 186 | 9053655 | ||
BC145108.1 | - | 2 | 186 | |||
BC150702.1 | - | 2 | 186 | 9087613 |