Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnmb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031169.4 | NP_112446.2 | 4 | 669 | ||
GenBank mRNA | AF020711.1 | - | 4 | 669 | ||
AJ001291.1 | - | 4 | 411 | |||
AK038987.1 | - | 4 | 397 | |||
BC013338.1 | - | 4 | 460 | 3988005 |