Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031186.3 | NP_112463.2 | 1 | 361 | ||
GenBank mRNA | AF221095.1 | - | 1 | 360 | ||
AK141945.1 | - | 2 | 510 | |||
BC079622.1 | - | 1 | 276 | 30535990 | ||
BC112404.1 | - | 1 | 273 | 40060424 |