Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164634.1 | NP_001158106.1 | 3 | 629 | ||
NM_001164635.1 | NP_001158107.1 | 3 | 537 | |||
NM_031194.5 | NP_112471.3 | 4 | 709 | |||
GenBank mRNA | AB079895.1 | - | 2 | 366 | ||
AF078869.1 | - | 3 | 484 | |||
AK031962.1 | - | 4 | 675 | |||
AK044336.1 | - | 3 | 506 | |||
AK157891.1 | - | 3 | 538 | |||
BC014762.1 | - | 3 | 515 | 4239544 | ||
BU615016.1 | - | 3 | 579 |