Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199271.1 | NP_001186200.1 | 2, 3 | 280 | ||
NM_031196.3 | NP_112473.1 | 2, 3 | 374 | |||
GenBank mRNA | AK002560.1 | - | 2, 3 | 291 | ||
AK050714.1 | - | 2, 3 | 404 | |||
AK079592.1 | - | 2, 3 | 335 | |||
AK085024.2 | - | 2, 3 | 320 | |||
AK088803.1 | - | 2, 3 | 337 | |||
AK138577.1 | - | 2, 3 | 335 | |||
BC015263.1 | - | 2, 3 | 292 | 4239690 | ||
BF786711.1 | - | 2, 3 | 291 | |||
L23755.1 | - | 1, 2 | 232 | |||
L36539.1 | - | 2, 3 | 287 | |||
U32469.1 | - | 2, 3 | 314 | |||
U66103.1 | - | 2, 3 | 327 |