Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031256.3 | NP_112546.1 | 2 | 412 | ||
GenBank mRNA | AF286163.1 | - | 2 | 201 | ||
BC012283.1 | - | 2 | 152 | |||
BC031110.1 | - | 3 | 220 | 4981486 | ||
BY730332.1 | - | 2 | 412 |