Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031257.3 | NP_112547.1 | 7 | 725 | ||
GenBank mRNA | AF286161.1 | - | 7 | 679 | ||
AF418551.1 | - | 6 | 591 | |||
AK032325.1 | - | 7 | 734 | |||
AK042324.1 | - | 7 | 699 | |||
AK078212.1 | - | 7 | 704 | |||
AK088395.1 | - | 7 | 667 | |||
AK153117.1 | - | 7 | 697 | |||
BC010215.1 | - | 7 | 646 | 3498236 |