Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tktl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031379.2 | NP_113556.2 | 4 | 417 | ||
GenBank mRNA | AF285571.1 | - | 4 | 392 | ||
AK029546.1 | - | 4 | 418 | |||
AK139841.1 | - | 4 | 417 | |||
BC113768.1 | - | 4 | 392 | 40058456 |