Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040085.2 | NP_001035174.1 | 3 | 885 | ||
NM_001289583.1 | NP_001276512.1 | 3 | 885 | |||
NM_001289584.1 | NP_001276513.1 | 3 | 885 | |||
NM_031394.3 | NP_113571.2 | 3 | 885 | |||
GenBank mRNA | AB057754.1 | - | 2 | 224 | ||
AB057760.1 | - | 2 | 224 | |||
AB057761.1 | - | 2 | 224 | |||
AB057762.1 | - | 2 | 224 | |||
BC052915.1 | - | 2 | 229 | |||
BC064769.1 | - | 2 | 217 | |||
BC138714.1 | - | 2 | 195 | 8861736 | ||
BC145398.1 | - | 2 | 195 | 9053956 |