Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031178.2 | NP_112455.2 | 2 | 1523 | ||
GenBank mRNA | AB045181.1 | - | 2 | 1523 | ||
AF314224.1 | - | 2 | 1502 | |||
AF348140.1 | - | 2 | 1456 |