Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031252.2 | NP_112542.1 | 4 | 833 | ||
GenBank mRNA | AF301619.1 | - | 4 | 833 | ||
BC019953.1 | - | 4 | 817 | 3708687 |