Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031874.4 | NP_114080.2 | 5 | 3258 | ||
GenBank mRNA | AF263365.1 | - | 5 | 3258 | ||
AK048395.1 | - | 5 | 3199 |