Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000351.5 | NP_000342.2 | 10 | 2503 | ||
NM_001320750.1 | NP_001307679.1 | 11 | 2727 | |||
NM_001320751.1 | NP_001307680.1 | 12 | 2721 | |||
NM_001320752.1 | NP_001307681.1 | 11 | 2567 | |||
NM_001320753.1 | NP_001307682.1 | 10 | 2345 | |||
NM_001320754.1 | NP_001307683.1 | 10 | 2493 | |||
GenBank mRNA | AL133910.1 | - | 1 | 538 | ||
M16505.1 | - | 10 | 2478 |