Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031880.3 | NP_114086.3 | 12 | 1873 | ||
GenBank mRNA | AF332512.1 | - | 10 | 1693 | ||
AK033440.1 | - | 7 | 1244 | |||
AK076220.1 | - | 12 | 1880 | |||
BC029623.1 | - | 11 | 1860 | 4039780 | ||
BC055303.1 | - | 11 | 1777 | 6439288 |