Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033759.2 | NP_001028931.1 | 10, 11 | 2516 | ||
NM_031997.4 | NP_114386.3 | 11, 12 | 2598 | |||
GenBank mRNA | AK129352.1 | - | 5, 6 | 817 | ||
BC019745.1 | - | 2, 3 | 239 | 3675225 | ||
BC076570.1 | - | 10, 11 | 2122 | 6830791 | ||
BC089353.1 | - | 10, 11 | 2498 | 6410437 | ||
BU058172.1 | - | 1, 2 | 29 | |||
CA752446.1 | - | 3, 4 | 456 |