Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033073.3 | NP_149064.1 | 2 | 442 | ||
GenBank mRNA | AF509888.1 | - | 2 | 419 | ||
AK002434.1 | - | 2 | 420 | |||
AK137217.1 | - | 420 | ||||
AK146914.1 | - | 2 | 420 | |||
BC010337.1 | - | 2 | 396 | |||
BC030403.1 | - | 2 | 453 | 5355662 |