Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D17H6S56E-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037701.1 | - | 2 | 614 | ||
AK075966.1 | - | 2 | 637 | |||
AK088284.1 | - | 3 | 684 | |||
AK088743.1 | - | 3 | 518 | |||
AK161228.1 | - | 2 | 612 | |||
BC027314.1 | - | 2 | 600 | 3985942 | ||
L78788.1 | - | 2 | 1005 |