Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033217.3 | NP_150086.2 | 2 | 244 | ||
GenBank mRNA | AF105292.1 | - | 2 | 102 | ||
AK037248.1 | - | 2 | 245 | |||
BB630380.1 | - | 2 | 245 | |||
BC038365.1 | - | 2 | 235 | 5367638 |