Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033269.4 | NP_150372.1 | 5 | 1069 | ||
GenBank mRNA | AK080950.1 | - | 5 | 1071 | ||
BC129892.1 | - | 5 | 564 | 40111498 | ||
BC129893.1 | - | 5 | 564 | 40111499 |
Set Membership: |
G-protein coupled receptor |